siRNA Details
| Gene Symbol: Cnot2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037848.3 | NP_001032937.1 | 5 | 937 | ||
| GenBank mRNA | AK172132.1 | - | 5 | 924 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnot2| Gene Symbol: Cnot2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037848.3 | NP_001032937.1 | 5 | 937 | ||
| GenBank mRNA | AK172132.1 | - | 5 | 924 | ||