siRNA Details
| Gene Symbol: Zfp606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026112.4 | NP_080388.2 | 7 | 3950 | ||
| GenBank mRNA | AK077352.1 | - | 3 | 3377 | ||
| AK143200.1 | - | 7 | 3915 | |||
| AK173278.1 | - | 8 | 3956 | |||
| BC052747.1 | - | 7 | 3870 | 6837952 | ||

Zfp606
Genomic Map