siRNA Details
| Gene Symbol: Zfp606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039951.2 | NP_001035040.1 | 4 | 972 | ||
| GenBank mRNA | AK032301.1 | - | 4 | 975 | ||
| AK039817.1 | - | 4 | 918 | |||
| AK165095.1 | - | 4 | 911 | |||
| BC110665.1 | - | 3 | 582 | |||

Zfp606
Genomic Map