siRNA Details
| Gene Symbol: Gm3238 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101630.1 | NP_001095100.1 | 1 | 383 | ||
| Gene Symbol: Gm19668 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205034.1 | NP_001191963.1 | 1 | 437 | ||
| Gene Symbol: Gm2696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205009.1 | NP_001191938.1 | 1 | 413 | ||

Gm3238
Genomic Map