siRNA Details
| Gene Symbol: Gm3238 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101630.1 | NP_001095100.1 | 1 | 628 | ||
| Gene Symbol: Gm3285 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101631.1 | NP_001095101.1 | 1 | 585 | ||

Gm3238
Genomic Map