siRNA Details
| Gene Symbol: Gm20831 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103152.1 | NP_001096622.1 | 1227 | |||
| GenBank mRNA | AK132684.1 | - | 1228 | |||
| AK161425.1 | - | 1194 | ||||
| Gene Symbol: Ssty1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009220.2 | NP_033246.2 | 3 | 1176 | ||
| GenBank mRNA | X05260.1 | - | 3 | 1176 | ||

Gm20831
Genomic Map