siRNA Details
| Gene Symbol: ZC3H7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017590.5 | NP_060060.3 | 8 | 887 | ||
| XM_011530016.2 | XP_011528318.1 | 8 | 868 | |||
| GenBank mRNA | AB028954.1 | - | 5 | 500 | ||
| AF188530.1 | - | 8 | 715 | |||
| AK225891.1 | - | 8 | 860 | |||
| AK315009.1 | - | 8 | 829 | |||
| BC098378.1 | - | 8 | 825 | |||
| BC152558.1 | - | 8 | 865 | 8860405 | ||

ZC3H7B
Genomic Map