siRNA Details
| Gene Symbol: Gm3285 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101631.1 | NP_001095101.1 | 1 | 66 | ||
| Gene Symbol: Gm10318 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162944.1 | NP_001156416.1 | 1 | 25 | ||

Gm3285
Genomic Map