siRNA Details
| Gene Symbol: Gm9268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105061.1 | NP_001098531.1 | 1 | 95 | ||
| Gene Symbol: Vmn2r62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105059.1 | NP_001098529.1 | 1 | 95 | ||
More Information
Set Membership: |
|

Gm9268
Genomic Map