siRNA Details
Gene Symbol: Gm9268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105061.1 | NP_001098531.1 | 4 | 682 |
Gene Symbol: Vmn2r62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105059.1 | NP_001098529.1 | 3 | 697 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
