siRNA Details
| Gene Symbol: Pde4dip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039376.2 | NP_001034465.2 | 38 | 6054 | ||
| NM_001289701.1 | NP_001276630.1 | 38 | 6105 | |||
| NM_001289702.1 | NP_001276631.1 | 37 | 5952 | |||
| GenBank mRNA | BC141185.1 | - | 38 | 6105 | 9055797 | |
| BC145504.1 | - | 37 | 5952 | 9054067 | ||
| BQ554472.1 | - | 3 | 374 | |||

Pde4dip
Genomic Map