siRNA Details
| Gene Symbol: Krtap4-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085547.2 | NP_001079016.1 | 1 | 434 | ||
| Gene Symbol: 2300003K06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195383.1 | NP_001182312.1 | 1 | 410 | ||

Krtap4-8
Genomic Map