siRNA Details
| Gene Symbol: Krtap4-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085547.2 | NP_001079016.1 | 1 | 478 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtap4-8| Gene Symbol: Krtap4-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085547.2 | NP_001079016.1 | 1 | 478 | ||