siRNA Details
| Gene Symbol: Rhox4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039689.1 | NP_001034778.1 | 2 | 531 | ||
| GenBank mRNA | AJ972667.1 | - | 2 | 531 | ||
| Gene Symbol: Rhox4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039688.3 | NP_001034777.1 | 2 | 519 | ||
| GenBank mRNA | AJ972665.1 | - | 2 | 519 | ||
| AK017380.1 | - | 2 | 434 | |||
| BC057925.1 | - | 2 | 434 | 5292192 | ||
| BC116370.1 | - | 2 | 467 | 40100097 | ||
| BC116371.1 | - | 2 | 466 | 40100098 | ||
| BC145382.1 | - | 2 | 420 | 9053940 | ||
| BC171959.1 | - | 2 | 420 | 9054245 | ||
| DQ058641.1 | - | 2 | 494 | |||
| Gene Symbol: Rhox4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021300.2 | NP_067275.1 | 2 | 514 | ||
| GenBank mRNA | AF265350.1 | - | 2 | 502 | ||
| AJ972666.1 | - | 2 | 514 | |||
| BC172126.1 | - | 2 | 461 | 9054796 | ||
| Gene Symbol: Rhox4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039695.1 | NP_001034784.1 | 387 | |||
| GenBank mRNA | AB221641.1 | - | 383 | |||
| AJ972668.1 | - | 387 | ||||
| BC147696.1 | - | 2 | 461 | 9056669 | ||
| Gene Symbol: Rhox4e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201236.3 | NP_957688.2 | 2 | 605 | ||
| GenBank mRNA | AJ972669.1 | - | 2 | 605 | ||
| BC094891.1 | - | 2 | 567 | 30019143 | ||
| BC172128.1 | - | 461 | 9054798 | |||
| Gene Symbol: Rhox4f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039696.1 | NP_001034785.1 | 2 | 557 | ||
| GenBank mRNA | AJ972670.1 | - | 2 | 557 | ||
| Gene Symbol: Rhox4g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039698.1 | NP_001034787.1 | 2 | 606 | ||
| GenBank mRNA | AJ972671.1 | - | 2 | 606 | ||

Rhox4c
Genomic Map