siRNA Details
| Gene Symbol: Gm3435 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001123372.1 | NP_001116844.1 | 1965 | |||
| GenBank mRNA | BC052508.1 | - | 962 | |||
| BC065801.1 | - | 953 | ||||
| Gene Symbol: 9030025P20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001123370.1 | NP_001116842.1 | 12 | 1569 | ||
| GenBank mRNA | AK161147.1 | - | 11 | 2364 | ||

Gm3435
Genomic Map