siRNA Details
| Gene Symbol: Gm3435 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001123372.1 | NP_001116844.1 | 1225 | |||
| GenBank mRNA | AK013428.1 | - | 1868 | |||
| AK078282.1 | - | 819 | ||||
| BC052508.1 | - | 222 | ||||
| BC065801.1 | - | 213 | ||||
| Gene Symbol: 9030025P20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001123370.1 | NP_001116842.1 | 8 | 829 | ||
| GenBank mRNA | AK042420.1 | - | 7 | 1762 | ||
| AK078855.1 | - | 8 | 817 | |||
| AK160486.1 | - | 8 | 881 | |||
| AK161147.1 | - | 7 | 1624 | |||
| Gene Symbol: Ermard | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039552.2 | NP_001034641.2 | 828 | |||
| GenBank mRNA | AK089867.1 | - | 1865 | |||
| BC150877.1 | - | 889 | 9087788 | |||

Gm3435
Genomic Map