siRNA Details
| Gene Symbol: Hist1h4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC115450.1 | - | 1 | 85 | 40099776 | |
| BC115451.1 | - | 1 | 78 | |||
| BC139809.1 | - | 1 | 85 | 40099779 | ||
| Gene Symbol: Hist1h4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178192.2 | NP_835499.1 | 1 | 177 | ||
| GenBank mRNA | AK016310.1 | - | 1 | 159 | ||
| BC119241.1 | - | 1 | 177 | 8733990 | ||
| BC119243.1 | - | 1 | 177 | 8733992 | ||
| Gene Symbol: Hist1h4f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175655.2 | NP_783586.1 | 1 | 85 | ||
| Gene Symbol: Hist1h4k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178211.2 | NP_835583.1 | 1 | 133 | ||
| GenBank mRNA | AK139521.1 | - | 1 | 120 | ||
| BC096767.1 | - | 100 | ||||
| BC117010.1 | - | 1 | 133 | 40126329 | ||
| BC117012.1 | - | 1 | 133 | 40126331 | ||
| BC119612.1 | - | 1 | 85 | 40061668 | ||

Hist1h4c
Genomic Map