siRNA Details
Gene Symbol: Col4a3bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF232932.1 | - | 18 | |||
AF232934.1 | - | 18 |
Gene Symbol: Mt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013602.3 | NP_038630.1 | 3 | 439 | ||
GenBank mRNA | AA050166.1 | - | 3 | 218 | ||
AK007983.1 | - | 3 | 288 | |||
AK011040.1 | - | 3 | 288 | |||
AK018727.1 | - | 3 | 302 | |||
AK154954.1 | - | 1 | 985 | |||
BC027262.1 | - | 2 | 530 | 3483861 | ||
BC036990.1 | - | 284 | 4990006 | |||
BC109369.1 | - | 798 |
