siRNA Details
| Gene Symbol: Jak3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010589.6 | NP_034719.2 | 25 | 3780 | ||
| GenBank mRNA | AK038268.1 | - | 24 | 3591 | ||
| AK088365.1 | - | 25 | 3629 | |||
| Gene Symbol: Insl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013564.7 | NP_038592.3 | 2 | 433 | ||
| GenBank mRNA | AK132857.1 | - | 2 | 434 | ||
| BC049540.1 | - | 2 | 433 | 6773462 | ||

Jak3
Genomic Map