siRNA Details
| Gene Symbol: Enho | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027147.1 | NP_081423.1 | 2 | 1224 | ||
| GenBank mRNA | AK009710.1 | - | 2 | 1224 | ||
| AK050496.1 | - | 2 | 1023 | |||
| AK207085.1 | - | 1 | 356 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Enho| Gene Symbol: Enho | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027147.1 | NP_081423.1 | 2 | 1224 | ||
| GenBank mRNA | AK009710.1 | - | 2 | 1224 | ||
| AK050496.1 | - | 2 | 1023 | |||
| AK207085.1 | - | 1 | 356 | |||