siRNA Details
| Gene Symbol: Gnas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077507.1 | NP_001070975.1 | 11 | 3384 | ||
| NM_001077510.2 | NP_001070978.1 | 11 | 1371 | |||
| NM_010309.3 | NP_034439.2 | 12 | 3426 | |||
| NM_022000.2 | NP_068840.2 | 13 | 2131 | |||
| NM_201616.1 | NP_963910.1 | 12 | 1413 | |||
| NM_201618.1 | NP_963912.1 | 12 | 3426 | |||
| GenBank mRNA | AF107848.1 | - | 13 | 2129 | ||
| AF116268.1 | - | 11 | 2313 | |||
| AK078037.1 | - | 12 | 1423 | |||
| AK132636.1 | - | 12 | 1514 | |||
| AK147051.1 | - | 11 | 1369 | |||
| AK159563.1 | - | 12 | 1414 | |||
| AK168996.1 | - | 12 | 1384 | |||
| AY519501.1 | - | 12 | 3426 | |||
| AY519502.1 | - | 12 | 3426 | |||
| AY519504.1 | - | 13 | 3521 | |||
| AY682719.1 | - | 13 | 1119 | |||
| BC013824.1 | - | 12 | 1225 | |||
| BC013840.1 | - | 12 | 1225 | |||
| BC018408.1 | - | 3 | 339 | |||
| BC034446.1 | - | 11 | 1465 | |||
| BC038067.1 | - | 13 | 1425 | |||
| BC048834.1 | - | 9 | 930 | |||
| BC061496.1 | - | 11 | 1355 | |||
| BC062654.1 | - | 13 | 3568 | |||
| BC080816.1 | - | 12 | 1402 | |||
| BC092055.1 | - | 12 | 1217 | |||
| BC106133.1 | - | 11 | 1177 | |||
| M13964.1 | - | 11 | 1171 | |||
| Y00703.1 | - | 12 | 1225 | |||

Gnas
Genomic Map