siRNA Details
Gene Symbol: Gnas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077507.1 | NP_001070975.1 | 11 | 3384 | ||
NM_001077510.2 | NP_001070978.1 | 11 | 1371 | |||
NM_010309.3 | NP_034439.2 | 12 | 3426 | |||
NM_022000.2 | NP_068840.2 | 13 | 2131 | |||
NM_201616.1 | NP_963910.1 | 12 | 1413 | |||
NM_201618.1 | NP_963912.1 | 12 | 3426 | |||
GenBank mRNA | AF107848.1 | - | 13 | 2129 | ||
AF116268.1 | - | 11 | 2313 | |||
AK078037.1 | - | 12 | 1423 | |||
AK132636.1 | - | 12 | 1514 | |||
AK147051.1 | - | 11 | 1369 | |||
AK159563.1 | - | 12 | 1414 | |||
AK168996.1 | - | 12 | 1384 | |||
AY519501.1 | - | 12 | 3426 | |||
AY519502.1 | - | 12 | 3426 | |||
AY519504.1 | - | 13 | 3521 | |||
AY682719.1 | - | 13 | 1119 | |||
BC013824.1 | - | 12 | 1225 | |||
BC013840.1 | - | 12 | 1225 | |||
BC018408.1 | - | 3 | 339 | |||
BC034446.1 | - | 11 | 1465 | |||
BC038067.1 | - | 13 | 1425 | |||
BC048834.1 | - | 9 | 930 | |||
BC061496.1 | - | 11 | 1355 | |||
BC062654.1 | - | 13 | 3568 | |||
BC080816.1 | - | 12 | 1402 | |||
BC092055.1 | - | 12 | 1217 | |||
BC106133.1 | - | 11 | 1177 | |||
M13964.1 | - | 11 | 1171 | |||
Y00703.1 | - | 12 | 1225 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
