siRNA Details
| Gene Symbol: Krtap10-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135991.1 | NP_001129463.1 | 1 | 764 | ||
| GenBank mRNA | AK020700.1 | - | 1 | 792 | ||
| BC115499.1 | - | 1 | 764 | 40100932 | ||
| BC115500.1 | - | 1 | 764 | 40100933 | ||

Krtap10-4
Genomic Map