siRNA Details
| Gene Symbol: Ece2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025462.2 | NP_079738.2 | 3 | 600 | ||
| GenBank mRNA | AK007407.1 | - | 3 | 609 | ||
| AK007791.1 | - | 3 | 602 | |||
| BC115541.2 | - | 3 | 602 | |||
| BC115542.1 | - | 2 | 1082 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ece2| Gene Symbol: Ece2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025462.2 | NP_079738.2 | 3 | 600 | ||
| GenBank mRNA | AK007407.1 | - | 3 | 609 | ||
| AK007791.1 | - | 3 | 602 | |||
| BC115541.2 | - | 3 | 602 | |||
| BC115542.1 | - | 2 | 1082 | |||