siRNA Details
Gene Symbol: Krtap5-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015808.1 | NP_056623.1 | 1 | 531 | ||
GenBank mRNA | BC104355.1 | - | 417 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Krtap5-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015808.1 | NP_056623.1 | 1 | 531 | ||
GenBank mRNA | BC104355.1 | - | 417 |