siRNA Details
| Gene Symbol: Hist1h4m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195421.1 | NP_001182350.1 | 1 | 219 | ||
| GenBank mRNA | BC139425.1 | - | 243 | 8862447 | ||
| BC139437.1 | - | 243 | 8862459 | |||
| Gene Symbol: Hist1h4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178192.2 | NP_835499.1 | 1 | 273 | ||
| GenBank mRNA | AK016310.1 | - | 1 | 255 | ||
| BC119241.1 | - | 1 | 273 | 8733990 | ||
| BC119243.1 | - | 1 | 273 | 8733992 | ||
| Gene Symbol: Hist1h4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178193.2 | NP_835500.1 | 1 | 265 | ||
| GenBank mRNA | BC132186.1 | - | 1 | 265 | 40130463 | |
| BC132212.1 | - | 1 | 265 | 40130489 | ||
| Gene Symbol: Hist1h4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178208.2 | NP_835515.1 | 1 | 190 | ||
| GenBank mRNA | BC115450.1 | - | 1 | 181 | 40099776 | |
| BC115451.1 | - | 1 | 174 | |||
| BC139809.1 | - | 1 | 181 | 40099779 | ||
| Gene Symbol: Hist1h4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175654.2 | NP_783585.1 | 1 | 181 | ||
| GenBank mRNA | BC119611.2 | - | 1 | 180 | ||
| Gene Symbol: Hist1h4f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175655.2 | NP_783586.1 | 1 | 181 | ||
| GenBank mRNA | BC115446.1 | - | 1 | 174 | ||
| Gene Symbol: Hist1h4h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153173.4 | NP_694813.1 | 1 | 252 | ||
| GenBank mRNA | AA028777.1 | - | 1 | 250 | ||
| AK007642.1 | - | 1 | 248 | |||
| BC028550.1 | - | 1 | 193 | 1398106 | ||
| BC058529.1 | - | 1 | 189 | 6817839 | ||
| BC087952.1 | - | 1 | 241 | 6787571 | ||
| Gene Symbol: Hist1h4i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175656.3 | NP_783587.1 | 1 | 220 | ||
| GenBank mRNA | AK011560.1 | - | 1 | 221 | ||
| BC019757.1 | - | 1 | 210 | |||
| BC052219.1 | - | 1 | 193 | |||
| BC092144.1 | - | 1 | 210 | 30612926 | ||
| BC125598.1 | - | 1 | 204 | 40130113 | ||
| BC125600.1 | - | 1 | 204 | 40130115 | ||
| Gene Symbol: Hist1h4j | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178210.1 | NP_835582.1 | 1 | 181 | ||
| GenBank mRNA | AK010085.1 | - | 1 | 217 | ||
| Gene Symbol: Hist1h4k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178211.2 | NP_835583.1 | 1 | 229 | ||
| GenBank mRNA | AK139521.1 | - | 1 | 216 | ||
| BC096767.1 | - | 196 | ||||
| BC117010.1 | - | 1 | 229 | 40126329 | ||
| BC117012.1 | - | 1 | 229 | 40126331 | ||
| BC119612.1 | - | 1 | 181 | 40061668 | ||
| Gene Symbol: Hist1h4n | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175657.2 | NP_783588.1 | 1 | 181 | ||
| GenBank mRNA | BC115448.2 | - | 1 | 181 | 40099764 | |
| BC152397.1 | - | 1 | 181 | 40099768 | ||
| Gene Symbol: Hist2h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033596.3 | NP_291074.1 | 1 | 194 | ||
| GenBank mRNA | BC051566.1 | - | 1 | 202 | ||
| BC057955.1 | - | 188 | 5359141 | |||
| BX520915.1 | - | 1 | 194 | |||
 Back To Top
 Back To TopMore Information
Other Species Targeted
 This siRNA also targets the following transcripts in Human: NM_003548, NM_001034077, BC093969, BC108260, CR542172, and others.
					            	This siRNA also targets the following transcripts in Human: NM_003548, NM_001034077, BC093969, BC108260, CR542172, and others.This siRNA also targets the following transcripts in Rat: NM_001271221, NM_022686, XM_008771773, XM_008771675, XM_008771676, and others.
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Hist1h4m
Hist1h4m Genomic Map
Genomic Map Back To Top
 Back To Top