siRNA Details
| Gene Symbol: Gm10220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134299.1 | NP_001127771.1 | 5 | 1942 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm10220| Gene Symbol: Gm10220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134299.1 | NP_001127771.1 | 5 | 1942 | ||