siRNA Details
| Gene Symbol: Inafm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136270.1 | NP_001129742.1 | 1 | 1236 | ||
| GenBank mRNA | AK008271.1 | - | 1 | 39 | ||
| AK011395.1 | - | 1 | 479 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Inafm1| Gene Symbol: Inafm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136270.1 | NP_001129742.1 | 1 | 1236 | ||
| GenBank mRNA | AK008271.1 | - | 1 | 39 | ||
| AK011395.1 | - | 1 | 479 | |||