siRNA Details
| Gene Symbol: KLHL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025010.4 | NP_079286.2 | 2 | 362 | ||
| XM_005265001.3 | XP_005265058.2 | 2 | 262 | |||
| XM_005265002.4 | XP_005265059.2 | 2 | 262 | |||
| XM_005265003.3 | XP_005265060.2 | 2 | 262 | |||
| XM_006713079.1 | XP_006713142.1 | 2 | 218 | |||
| GenBank mRNA | AB062478.1 | - | 2 | 534 | ||
| AI356423.1 | - | 2 | 362 | |||
| AK023765.1 | - | 2 | 256 | |||
| AK291477.1 | - | 2 | 256 | |||
| BC032620.1 | - | 2 | 174 | 5578173 | ||
| BX537953.1 | - | 2 | 253 | |||

KLHL18
Genomic Map