siRNA Details
Gene Symbol: Mup1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163010.1 | NP_001156482.1 | 4 | 455 | ||
NM_001163011.1 | NP_001156483.1 | 4 | 455 | |||
NM_031188.2 | NP_112465.2 | 4 | 455 | |||
GenBank mRNA | BC012221.1 | - | 4 | 401 | 4195753 | |
BC013649.1 | - | 389 | 4195401 | |||
BC037152.1 | - | 279 | 4161709 | |||
BC091744.1 | - | 392 | 5102271 | |||
BC091775.1 | - | 4 | 402 | 5137341 | ||
BC092254.1 | - | 389 | 5100458 | |||
BC093515.1 | - | 389 | ||||
BC099597.1 | - | 392 | 947916 | |||
BC100586.1 | - | 409 | 6769126 | |||
BC106092.1 | - | 4 | 392 | 5098340 | ||
M16355.1 | - | 4 | 455 | |||
M27609.1 | - | 257 | ||||
M28649.1 | - | 402 | ||||
X03524.1 | - | 257 | ||||
X04115.1 | - | 213 |
Gene Symbol: Gm2083 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134644.1 | NP_001128116.1 | 4 | 409 |
Gene Symbol: LOC100048884 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199333.1 | NP_001186262.1 | 4 | 416 | ||
GenBank mRNA | BC055353.1 | - | 391 |
Gene Symbol: Mup10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122647.1 | NP_001116119.1 | 4 | 411 | ||
GenBank mRNA | AJ309921.1 | - | 344 | |||
AK011413.1 | - | 2 | 463 | |||
CB948918.1 | - | 320 | ||||
EU882231.1 | - | 4 | 344 |
Gene Symbol: Mup11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164526.1 | NP_001157998.1 | 442 | |||
GenBank mRNA | AK013972.1 | - | 4 | 419 |
Gene Symbol: Mup12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199995.1 | NP_001186924.1 | 4 | 410 |
Gene Symbol: Mup13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134674.1 | NP_001128146.1 | 4 | 470 |
Gene Symbol: Mup14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199999.1 | NP_001186928.1 | 4 | 453 |
Gene Symbol: Mup15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200004.1 | NP_001186933.1 | 4 | 416 |
Gene Symbol: Mup16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199936.1 | NP_001186865.1 | 4 | 410 |
Gene Symbol: Mup17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200006.1 | NP_001186935.1 | 4 | 416 |
Gene Symbol: Mup19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135127.2 | NP_001128599.1 | 4 | 410 | ||
GenBank mRNA | AK013259.1 | - | 4 | 410 | ||
EU882232.1 | - | 4 | 338 |
Gene Symbol: Mup2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045550.2 | NP_001039015.1 | 4 | 414 | ||
NM_001286096.1 | NP_001273025.1 | 3 | 280 | |||
NM_008647.4 | NP_032673.3 | 4 | 414 | |||
GenBank mRNA | BC012259.1 | - | 403 | 4196403 | ||
BC059097.1 | - | 389 | 4159984 | |||
CB593503.1 | - | 412 | ||||
M16356.1 | - | 402 |
Gene Symbol: Mup7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134675.1 | NP_001128147.1 | 4 | 519 | ||
GenBank mRNA | EU882230.1 | - | 4 | 344 |
Gene Symbol: Mup8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134676.1 | NP_001128148.1 | 4 | 474 |
Gene Symbol: Mup9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281979.1 | NP_001268908.1 | 4 | 410 | ||
GenBank mRNA | BI219655.1 | - | 4 | 391 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC094452.1 | - | 390 | Chr. - - [-] on Build |
