siRNA Details
| Gene Symbol: RGD1560394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106014.1 | NP_001099484.1 | 3 | 623 | ||
| XM_006251193.2 | XP_006251255.1 | 4 | 467 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1560394| Gene Symbol: RGD1560394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106014.1 | NP_001099484.1 | 3 | 623 | ||
| XM_006251193.2 | XP_006251255.1 | 4 | 467 | |||