siRNA Details
| Gene Symbol: CLUH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015229.3 | NP_056044.3 | 16 | 2802 | ||
| XM_011523768.2 | XP_011522070.1 | 15 | 2733 | |||
| GenBank mRNA | AB014564.1 | - | 12 | 2191 | ||
| AK023003.1 | - | 8 | 1652 | |||
| AK125717.1 | - | 16 | 2821 | |||
| AK308854.1 | - | 1 | 74 | |||
| BC063786.1 | - | 10 | 1884 | |||
| CR627449.1 | - | 14 | 2540 | |||

CLUH
Genomic Map