siRNA Details
| Gene Symbol: Mars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127659.1 | NP_001121131.1 | 15 | 1933 | ||
| XM_006241451.2 | XP_006241513.1 | 15 | 2046 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mars| Gene Symbol: Mars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127659.1 | NP_001121131.1 | 15 | 1933 | ||
| XM_006241451.2 | XP_006241513.1 | 15 | 2046 | |||