siRNA Details
| Gene Symbol: LOC689927 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109555.1 | NP_001103025.1 | 869 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC689927| Gene Symbol: LOC689927 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109555.1 | NP_001103025.1 | 869 | |||