siRNA Details
| Gene Symbol: Vom2r72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099517.1 | NP_001092987.1 | 1140 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vom2r72| Gene Symbol: Vom2r72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099517.1 | NP_001092987.1 | 1140 | |||