siRNA Details
Gene Symbol: RGD1563667 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100985.1 | NP_001094455.1 | 2 | 506 | ||
GenBank mRNA | AY902367.1 | - | 2 | 506 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1563667 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100985.1 | NP_001094455.1 | 2 | 506 | ||
GenBank mRNA | AY902367.1 | - | 2 | 506 |