siRNA Details
| Gene Symbol: Hdlbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172039.2 | NP_742036.2 | 24 | 3420 | ||
| XM_006245548.1 | XP_006245610.1 | 24 | 3460 | |||
| GenBank mRNA | BC105748.1 | - | 24 | 3420 | 5598374 | |
| U90725.1 | - | 24 | 3420 | |||

Hdlbp
Genomic Map