siRNA Details
| Gene Symbol: NUP160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015231.2 | NP_056046.1 | 8 | 1257 | ||
| NR_134636.1 | 8 | 1257 | ||||
| GenBank mRNA | AK302396.1 | - | 7 | 729 | ||
| AK304308.1 | - | 8 | 1043 | |||
| BC125227.1 | - | 8 | 1180 | 40124592 | ||
| BC125228.1 | - | 8 | 1180 | 40124593 | ||
| BX647634.1 | - | 8 | 1042 | |||
| D83781.1 | - | 8 | 1026 | |||

NUP160
Genomic Map