siRNA Details
| Gene Symbol: Degs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053323.2 | NP_445775.2 | 2 | 553 | ||
| XM_006250403.2 | XP_006250465.1 | 2 | 848 | |||
| XM_006250404.2 | XP_006250466.1 | 2 | 794 | |||
| GenBank mRNA | AJ938081.1 | - | 2 | 506 | ||
| AY036902.1 | - | 2 | 535 | |||
| BC083727.1 | - | 2 | 553 | 7191263 | ||
| FQ225269.1 | - | 2 | 561 | |||

Degs1
Genomic Map