siRNA Details
| Gene Symbol: Degs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053323.2 | NP_445775.2 | 3 | 893 | ||
| XM_006250403.2 | XP_006250465.1 | 3 | 1188 | |||
| XM_006250404.2 | XP_006250466.1 | 3 | 1134 | |||
| GenBank mRNA | AJ938081.1 | - | 3 | 846 | ||
| AY036902.1 | - | 3 | 875 | |||
| BC083727.1 | - | 3 | 893 | 7191263 | ||
| FQ225269.1 | - | 3 | 901 | |||

Degs1
Genomic Map