siRNA Details
Gene Symbol: Olr837 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000897.1 | NP_001000897.1 | 1 | 639 | ||
GenBank mRNA | AF091565.1 | - | 1 | 447 |
Gene Symbol: LOC103692344 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763684.1 | XP_008761906.1 | 1032 |
