siRNA Details
| Gene Symbol: Gltscr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106226.2 | NP_001099696.2 | 8, 9 | 3211 | ||
| XM_006228343.2 | XP_006228405.1 | 10, 11 | 3558 | |||
| XM_006228344.2 | XP_006228406.1 | 9, 10 | 3492 | |||
| XM_006228345.2 | XP_006228407.1 | 9, 10 | 3381 | |||

Gltscr1
Genomic Map