siRNA Details
| Gene Symbol: LOC100151767 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127503.1 | NP_001120975.1 | 4 | 209 | ||
| XM_006243024.2 | XP_006243086.1 | 4 | 222 | |||
| XM_006243025.2 | XP_006243087.1 | 4 | 741 | |||
| XM_006243026.2 | XP_006243088.1 | 4 | 741 | |||
| GenBank mRNA | BC161965.1 | - | 4 | 209 | 9028305 | |

LOC100151767
Genomic Map