siRNA Details
| Gene Symbol: Kcng2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107372.1 | NP_001100842.1 | 2 | 256 | ||
| XM_006254947.2 | XP_006255009.1 | 2 | 1630 | |||
| XM_006254948.2 | XP_006255010.1 | 2 | 800 | |||
| XM_006254950.2 | XP_006255012.1 | 2 | 151 | |||
| XM_008772136.1 | XP_008770358.1 | 2 | 767 | |||
| XM_008772137.1 | XP_008770359.1 | 2 | 195 | |||
| GenBank mRNA | AJ011020.1 | - | 1 | 122 | ||

Kcng2
Genomic Map