siRNA Details
| Gene Symbol: Rpl19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031103.1 | NP_112365.1 | 4, 5 | 377 | ||
| GenBank mRNA | BC058135.1 | - | 4, 5 | 377 | 6918377 | |
| FQ209882.1 | - | 2, 3 | 206 | |||
| FQ211447.1 | - | 2, 3 | 206 | |||
| FQ221779.1 | - | 2, 3 | 206 | |||
| FQ221820.1 | - | 2, 3 | 206 | |||
| FQ222282.1 | - | 4, 5 | 377 | |||
| J02650.1 | - | 4, 5 | 377 | |||

Rpl19
Genomic Map