siRNA Details
| Gene Symbol: LOC103689994 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751965.3 | XP_003752013.1 | 1920 | |||
| Gene Symbol: Rsph4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751961.3 | XP_003752009.1 | 4 | 1920 | ||

LOC103689994
Genomic Map