siRNA Details
| Gene Symbol: LOC497796 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111310.1 | NP_001104780.1 | 738 | |||
| XM_006237126.2 | XP_006237188.1 | 6 | 788 | |||
| GenBank mRNA | BC093399.1 | - | 738 | 7389533 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC497796| Gene Symbol: LOC497796 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111310.1 | NP_001104780.1 | 738 | |||
| XM_006237126.2 | XP_006237188.1 | 6 | 788 | |||
| GenBank mRNA | BC093399.1 | - | 738 | 7389533 | ||