siRNA Details
| Gene Symbol: Rap2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053741.1 | NP_446193.1 | 1 | 422 | ||
| GenBank mRNA | AY037826.1 | - | 1 | 422 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rap2a| Gene Symbol: Rap2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053741.1 | NP_446193.1 | 1 | 422 | ||
| GenBank mRNA | AY037826.1 | - | 1 | 422 | ||