siRNA Details
| Gene Symbol: Enpp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019370.2 | NP_062243.2 | 18 | 1721 | ||
| GenBank mRNA | BC097326.1 | - | 18 | 1721 | 7458431 | |
| FQ218696.1 | - | 1 | 106 | |||
| U78788.1 | - | 18 | 1724 | |||
| Z47987.1 | - | 18 | 1705 | |||

Enpp3
Genomic Map