siRNA Details
| Gene Symbol: Spatc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115026.1 | NP_001108498.1 | 1 | 188 | ||
| GenBank mRNA | BC083658.1 | - | 1 | 177 | ||
| BC103732.1 | - | 1 | 176 | |||
| BC158623.1 | - | 1 | 183 | 7454231 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spatc1| Gene Symbol: Spatc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115026.1 | NP_001108498.1 | 1 | 188 | ||
| GenBank mRNA | BC083658.1 | - | 1 | 177 | ||
| BC103732.1 | - | 1 | 176 | |||
| BC158623.1 | - | 1 | 183 | 7454231 | ||